Assay Details
| Gene Symbol: Adgf-D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079621.3 | NP_524345.2 | 3 - 4 | 806 | 90 | |
| GenBank mRNA | AF337553.1 | - | 3 - 4 | 787 | 90 | |
| BT010219.1 | - | 3 - 4 | 861 | 90 | ||
More Information
Set Membership: |
|

Adgf-D
Genomic Map