Assay Details
| Gene Symbol: Scp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079657.4 | NP_524381.1 | 4 - 5 | 437 | 66 | |
| GenBank mRNA | AF014952.1 | - | 3 - 4 | 260 | 66 | |
| AF093240.1 | - | 4 - 5 | 406 | 66 | ||
| AY089680.1 | - | 4 - 5 | 437 | 66 | ||
More Information
Set Membership: |
|

Scp2
Genomic Map