Assay Details
Gene Symbol: cdi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260262.2 | NP_001247191.1 | 2 - 3 | 1477 | 73 | |
NM_079677.4 | NP_524401.2 | 2 - 3 | 1477 | 73 | ||
GenBank mRNA | BT004492.1 | - | 1 - 2 | 523 | 73 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
