Assay Details
Gene Symbol: Dys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043256.2 | NP_001036721.1 | 5 - 6 | 2039 | 82 | |
NM_001043257.2 | NP_001036722.1 | 21 - 22 | 7886 | 82 | ||
NM_001043258.2 | NP_001036723.1 | 21 - 22 | 7886 | 82 | ||
NM_001043259.2 | NP_001036724.1 | 19 - 20 | 6818 | 82 | ||
NM_001043261.2 | NP_001036726.1 | 20 - 21 | 7599 | 82 | ||
NM_001043263.2 | NP_001036728.1 | 20 - 21 | 7813 | 82 | ||
NM_001275801.1 | NP_001262730.1 | 18 - 19 | 6703 | 82 | ||
NM_001275803.1 | NP_001262732.1 | 19 - 20 | 6818 | 82 | ||
NM_001275804.1 | NP_001262733.1 | 9 - 10 | 1361 | 82 | ||
GenBank mRNA | AF277386.1 | - | 18 - 19 | 6387 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
