Assay Details
Gene Symbol: Dys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043256.2 | NP_001036721.1 | 8 - 9 | 2444 | 55 | |
NM_001043257.2 | NP_001036722.1 | 24 - 25 | 8291 | 55 | ||
NM_001043258.2 | NP_001036723.1 | 24 - 25 | 8291 | 55 | ||
NM_001043259.2 | NP_001036724.1 | 22 - 23 | 7223 | 55 | ||
NM_001043260.2 | NP_001036725.1 | 4 - 5 | 3227 | 55 | ||
NM_001043261.2 | NP_001036726.1 | 23 - 24 | 8004 | 55 | ||
NM_001043263.2 | NP_001036728.1 | 23 - 24 | 8218 | 55 | ||
NM_001275801.1 | NP_001262730.1 | 21 - 22 | 7108 | 55 | ||
NM_001275803.1 | NP_001262732.1 | 22 - 23 | 7223 | 55 | ||
NM_001275804.1 | NP_001262733.1 | 12 - 13 | 1766 | 55 | ||
GenBank mRNA | AF277386.1 | - | 21 - 22 | 6792 | 55 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
