Assay Details
Gene Symbol: Dys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043256.2 | NP_001036721.1 | 10 - 11 | 2729 | 79 | |
NM_001043257.2 | NP_001036722.1 | 26 - 27 | 8576 | 79 | ||
NM_001043258.2 | NP_001036723.1 | 26 - 27 | 8576 | 79 | ||
NM_001043259.2 | NP_001036724.1 | 24 - 25 | 7508 | 79 | ||
NM_001043260.2 | NP_001036725.1 | 6 - 7 | 3512 | 79 | ||
NM_001043261.2 | NP_001036726.1 | 25 - 26 | 8289 | 79 | ||
NM_001043263.2 | NP_001036728.1 | 25 - 26 | 8503 | 79 | ||
GenBank mRNA | AF277386.1 | - | 23 - 24 | 7077 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
