Assay Details
Gene Symbol: Dys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043256.2 | NP_001036721.1 | 14 - 15 | 3687 | 61 | |
NM_001043257.2 | NP_001036722.1 | 30 - 31 | 9534 | 61 | ||
NM_001043258.2 | NP_001036723.1 | 30 - 31 | 9534 | 61 | ||
NM_001043259.2 | NP_001036724.1 | 28 - 29 | 8466 | 61 | ||
NM_001043260.2 | NP_001036725.1 | 10 - 11 | 4470 | 61 | ||
NM_001043261.2 | NP_001036726.1 | 29 - 30 | 9247 | 61 | ||
NM_001043262.2 | NP_001036727.1 | 6 - 7 | 2694 | 61 | ||
NM_001043263.2 | NP_001036728.1 | 29 - 30 | 9461 | 61 | ||
NM_001275801.1 | NP_001262730.1 | 28 - 29 | 8402 | 61 | ||
NM_001275802.1 | NP_001262731.1 | 6 - 7 | 2293 | 61 | ||
NM_001275803.1 | NP_001262732.1 | 29 - 30 | 8517 | 61 | ||
NM_001275804.1 | NP_001262733.1 | 19 - 20 | 3060 | 61 | ||
GenBank mRNA | AF277386.1 | - | 27 - 28 | 8035 | 61 | |
BT010020.1 | - | 6 - 7 | 2399 | 61 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
