Assay Details
Gene Symbol: Dys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043256.2 | NP_001036721.1 | 15 - 16 | 4072 | 69 | |
NM_001043257.2 | NP_001036722.1 | 31 - 32 | 9919 | 69 | ||
NM_001043258.2 | NP_001036723.1 | 31 - 32 | 9919 | 69 | ||
NM_001043259.2 | NP_001036724.1 | 29 - 30 | 8851 | 69 | ||
NM_001043260.2 | NP_001036725.1 | 11 - 12 | 4855 | 69 | ||
NM_001043261.2 | NP_001036726.1 | 30 - 31 | 9632 | 69 | ||
NM_001043262.2 | NP_001036727.1 | 7 - 8 | 3079 | 69 | ||
NM_001043263.2 | NP_001036728.1 | 30 - 31 | 9846 | 69 | ||
NM_001275801.1 | NP_001262730.1 | 29 - 30 | 8787 | 69 | ||
NM_001275802.1 | NP_001262731.1 | 7 - 8 | 2678 | 69 | ||
NM_001275803.1 | NP_001262732.1 | 30 - 31 | 8902 | 69 | ||
NM_001275804.1 | NP_001262733.1 | 20 - 21 | 3445 | 69 | ||
GenBank mRNA | AF277386.1 | - | 28 - 29 | 8420 | 69 | |
BT010020.1 | - | 7 - 8 | 2784 | 69 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
