Assay Details
Gene Symbol: Dys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043256.2 | NP_001036721.1 | 16 - 17 | 4190 | 54 | |
NM_001043257.2 | NP_001036722.1 | 32 - 33 | 10037 | 54 | ||
NM_001043258.2 | NP_001036723.1 | 32 - 33 | 10037 | 54 | ||
NM_001043259.2 | NP_001036724.1 | 30 - 31 | 8969 | 54 | ||
NM_001043260.2 | NP_001036725.1 | 12 - 13 | 4973 | 54 | ||
NM_001043261.2 | NP_001036726.1 | 31 - 32 | 9750 | 54 | ||
NM_001043262.2 | NP_001036727.1 | 8 - 9 | 3197 | 54 | ||
NM_001043263.2 | NP_001036728.1 | 31 - 32 | 9964 | 54 | ||
NM_001275801.1 | NP_001262730.1 | 30 - 31 | 8905 | 54 | ||
NM_001275802.1 | NP_001262731.1 | 8 - 9 | 2796 | 54 | ||
NM_001275803.1 | NP_001262732.1 | 31 - 32 | 9020 | 54 | ||
NM_001275804.1 | NP_001262733.1 | 21 - 22 | 3563 | 54 | ||
GenBank mRNA | BT010020.1 | - | 8 - 9 | 2902 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
