Assay Details
Gene Symbol: Dys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043256.2 | NP_001036721.1 | 17 - 18 | 4620 | 102 | |
NM_001043257.2 | NP_001036722.1 | 33 - 34 | 10467 | 102 | ||
NM_001043258.2 | NP_001036723.1 | 33 - 34 | 10467 | 102 | ||
NM_001043259.2 | NP_001036724.1 | 31 - 32 | 9399 | 102 | ||
NM_001043260.2 | NP_001036725.1 | 13 - 14 | 5403 | 102 | ||
NM_001043261.2 | NP_001036726.1 | 32 - 33 | 10180 | 102 | ||
NM_001043262.2 | NP_001036727.1 | 9 - 10 | 3627 | 102 | ||
NM_001043263.2 | NP_001036728.1 | 32 - 33 | 10394 | 102 | ||
NM_001275801.1 | NP_001262730.1 | 31 - 32 | 9335 | 102 | ||
NM_001275802.1 | NP_001262731.1 | 9 - 10 | 3226 | 102 | ||
NM_001275803.1 | NP_001262732.1 | 32 - 33 | 9450 | 102 | ||
NM_001275804.1 | NP_001262733.1 | 22 - 23 | 3993 | 102 | ||
GenBank mRNA | AF277386.1 | - | 30 - 31 | 8968 | 102 | |
BT010020.1 | - | 9 - 10 | 3332 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
