Assay Details
Gene Symbol: Dys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043257.2 | NP_001036722.1 | 5 - 6 | 4389 | 124 | |
NM_001043258.2 | NP_001036723.1 | 5 - 6 | 4389 | 124 | ||
NM_001043259.2 | NP_001036724.1 | 5 - 6 | 4431 | 124 | ||
NM_001043261.2 | NP_001036726.1 | 4 - 5 | 4102 | 124 | ||
NM_001043263.2 | NP_001036728.1 | 4 - 5 | 4316 | 124 | ||
NM_001275801.1 | NP_001262730.1 | 4 - 5 | 4316 | 124 | ||
NM_001275803.1 | NP_001262732.1 | 5 - 6 | 4431 | 124 | ||
GenBank mRNA | AF277386.1 | - | 4 - 5 | 4000 | 124 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
