Assay Details
| Gene Symbol: Calx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079699.4 | NP_524423.2 | 3 - 4 | 3210 | 103 | |
| NM_169940.4 | NP_732576.1 | 4 - 5 | 2613 | 103 | ||
| NM_169941.2 | NP_732577.1 | 3 - 4 | 2455 | 103 | ||
| GenBank mRNA | AY118680.1 | - | 4 - 5 | 2559 | 103 | |
| BT011420.1 | - | 4 - 5 | 2556 | 103 | ||
More Information
Set Membership: |
|

Calx
Genomic Map