Assay Details
| Gene Symbol: Cortactin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260293.2 | NP_001247222.1 | 2 - 3 | 366 | 146 | |
| NM_001260294.1 | NP_001247223.1 | 3 - 4 | 338 | 146 | ||
| NM_001260295.1 | NP_001247224.1 | 3 - 4 | 335 | 146 | ||
| NM_079702.3 | NP_524426.2 | 2 - 3 | 363 | 146 | ||
| GenBank mRNA | AY051774.1 | - | 2 - 3 | 311 | 146 | |
More Information
Set Membership: |
|

Cortactin
Genomic Map