Assay Details
Gene Symbol: InR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144621.3 | NP_001138093.1 | 5 - 6 | 2710 | 114 | |
NM_001144622.2 | NP_001138094.1 | 4 - 5 | 2011 | 114 | ||
NM_001144623.2 | NP_001138095.1 | 4 - 5 | 2077 | 114 | ||
NM_079712.6 | NP_524436.2 | 5 - 6 | 2720 | 114 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
