Assay Details
Gene Symbol: InR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144621.3 | NP_001138093.1 | 8 - 9 | 3531 | 72 | |
NM_001144622.2 | NP_001138094.1 | 7 - 8 | 2832 | 72 | ||
NM_001144623.2 | NP_001138095.1 | 7 - 8 | 2898 | 72 | ||
NM_079712.6 | NP_524436.2 | 8 - 9 | 3541 | 72 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
