Assay Details
| Gene Symbol: Mitofilin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275889.1 | NP_001262818.1 | 3 - 4 | 887 | 67 | |
| NM_001275890.1 | NP_001262819.1 | 3 - 4 | 887 | 67 | ||
| NM_079720.3 | NP_524444.3 | 3 - 4 | 1148 | 67 | ||
| GenBank mRNA | AF145623.1 | - | 3 - 4 | 850 | 67 | |
More Information
Set Membership: |
|

Mitofilin
Genomic Map