Assay Details
| Gene Symbol: Mitofilin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275889.1 | NP_001262818.1 | 5 - 6 | 1741 | 66 | |
| NM_079720.3 | NP_524444.3 | 5 - 6 | 2002 | 66 | ||
| GenBank mRNA | AF145623.1 | - | 5 - 6 | 1704 | 66 | |
More Information
Set Membership: |
|

Mitofilin
Genomic Map