Assay Details
Gene Symbol: Nrx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170216.3 | NP_001163687.1 | 6 - 7 | 2864 | 70 | |
NM_001275908.2 | NP_001262837.1 | 5 - 6 | 3202 | 70 | ||
NM_001275909.2 | NP_001262838.1 | 5 - 6 | 3202 | 70 | ||
NM_001275910.2 | NP_001262839.1 | 5 - 6 | 3202 | 70 | ||
NM_001275911.2 | NP_001262840.1 | 5 - 6 | 3202 | 70 | ||
NM_001275912.2 | NP_001262841.1 | 5 - 6 | 3499 | 70 | ||
NM_001275913.2 | NP_001262842.1 | 5 - 6 | 3202 | 70 | ||
NM_001300538.1 | NP_001287467.1 | 5 - 6 | 3202 | 70 | ||
NM_079725.5 | NP_524449.1 | 6 - 7 | 3302 | 70 | ||
GenBank mRNA | BT023898.1 | - | 6 - 7 | 2840 | 70 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
