Assay Details
Gene Symbol: CCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079726.4 | NP_524450.2 | 1 - 2 | 182 | 100 | |
NM_170016.2 | NP_732748.1 | 2 - 3 | 118 | 100 | ||
GenBank mRNA | AY118416.1 | - | 1 - 2 | 178 | 100 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
