Assay Details
Gene Symbol: CCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079726.4 | NP_524450.2 | 3 - 4 | 1222 | 87 | |
NM_170016.2 | NP_732748.1 | 4 - 5 | 1158 | 87 | ||
GenBank mRNA | AY118416.1 | - | 3 - 4 | 1218 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
