Assay Details
Gene Symbol: pnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079737.3 | NP_524461.2 | 5 - 6 | 1533 | 63 | |
NM_170651.4 | NP_732857.1 | 3 - 4 | 977 | 63 | ||
GenBank mRNA | BT058033.1 | - | 5 - 6 | 1529 | 63 | |
BT133301.1 | - | 4 - 5 | 935 | 63 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
