Assay Details
Gene Symbol: MP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144530.2 | NP_001138002.1 | 3 - 4 | 559 | 60 | |
NM_001316492.1 | NP_001303421.1 | 3 - 4 | 562 | 60 | ||
NM_141193.4 | NP_649450.3 | 3 - 4 | 532 | 60 | ||
GenBank mRNA | AF145611.1 | - | 3 - 4 | 527 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
