Assay Details
| Gene Symbol: Skp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164340.4 | NP_730815.2 | 3 - 4 | 1397 | 64 | |
| NM_164341.4 | NP_730816.3 | 3 - 4 | 1397 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Skp2| Gene Symbol: Skp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164340.4 | NP_730815.2 | 3 - 4 | 1397 | 64 | |
| NM_164341.4 | NP_730816.3 | 3 - 4 | 1397 | 64 | ||
Set Membership: |
|