Assay Details
| Gene Symbol: Tim17b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141269.3 | NP_649526.2 | 2 - 3 | 569 | 62 | |
| GenBank mRNA | AY113269.1 | - | 2 - 3 | 569 | 62 | |
| BT029703.1 | - | 1289 | 62 | |||
More Information
Set Membership: |
|

Tim17b1
Genomic Map