Assay Details
Gene Symbol: Prosbeta7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141272.1 | NP_649529.1 | 2 - 3 | 947 | 63 | |
NM_169049.2 | NP_730922.1 | 2 - 3 | 758 | 63 | ||
GenBank mRNA | AY118936.1 | - | 2 - 3 | 715 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
