Assay Details
| Gene Symbol: RpL35A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141282.3 | NP_649539.1 | 4 - 5 | 525 | 95 | |
| GenBank mRNA | AY071136.1 | - | 4 - 5 | 521 | 95 | |
| AY071732.1 | - | 4 - 5 | 523 | 95 | ||
More Information
Set Membership: |
|

RpL35A
Genomic Map