Assay Details
Gene Symbol: Atg17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141316.2 | NP_649573.2 | 5 - 6 | 4391 | 77 | |
NM_206434.2 | NP_996156.1 | 4 - 5 | 4370 | 77 | ||
GenBank mRNA | BT133015.1 | - | 4 - 5 | 4368 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
