Assay Details
Gene Symbol: wa-cup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141464.2 | NP_649721.2 | 1 - 2 | 383 | 57 | |
GenBank mRNA | AY119089.1 | - | 1 - 2 | 383 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
