Assay Details
Gene Symbol: wa-cup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141464.2 | NP_649721.2 | 6 - 7 | 1389 | 65 | |
GenBank mRNA | AY119089.1 | - | 6 - 7 | 1389 | 65 |

More Information
Set Membership: |
![]() ![]() |
