Assay Details
| Gene Symbol: CD98hc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275425.1 | NP_001262354.2 | 6 - 7 | 1739 | 63 | |
| NM_001275426.1 | NP_001262355.1 | 6 - 7 | 1812 | 63 | ||
| NM_141495.3 | NP_649752.2 | 3 - 4 | 1662 | 63 | ||
| GenBank mRNA | BT044083.1 | - | 3 - 4 | 1659 | 63 | |
More Information
Set Membership: |
|

CD98hc
Genomic Map