Assay Details
Gene Symbol: Ada2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275438.1 | NP_001262367.1 | 2 - 3 | 1080 | 65 | |
NM_141516.3 | NP_649773.1 | 2 - 3 | 1080 | 65 | ||
GenBank mRNA | AY058546.1 | - | 2 - 3 | 1073 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
