Assay Details
| Gene Symbol: CG7483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141531.3 | NP_649788.2 | 3 - 4 | 825 | 100 | |
| GenBank mRNA | BT133445.1 | - | 3 - 4 | 819 | 100 | |
More Information
Set Membership: |
|

CG7483
Genomic Map