Assay Details
| Gene Symbol: SLIRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141551.2 | NP_649808.1 | 1 | 80 | 62 | |
| GenBank mRNA | BT031072.1 | - | 1 | 80 | 62 | |
| BT032713.1 | - | 1 | 80 | 62 | ||
| KX531293.1 | - | 1 | 47 | 62 | ||
More Information
Set Membership: |
|

SLIRP2
Genomic Map