Assay Details
Gene Symbol: Prosbeta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141601.2 | NP_649858.1 | 1 | 857 | 137 | |
GenBank mRNA | AY095029.1 | - | 1 | 857 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
