Assay Details
| Gene Symbol: Vps16A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141620.3 | NP_649877.1 | 5 - 6 | 1435 | 76 | |
| GenBank mRNA | AY071611.1 | - | 5 - 6 | 1422 | 76 | |
More Information
Set Membership: |
|

Vps16A
Genomic Map