Assay Details
Gene Symbol: Vps16A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141620.3 | NP_649877.1 | 8 - 9 | 2460 | 63 | |
GenBank mRNA | AY071611.1 | - | 8 - 9 | 2447 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
