Assay Details
Gene Symbol: RnpS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141646.2 | NP_649903.1 | 2 - 3 | 314 | 89 | |
GenBank mRNA | AJ459409.1 | - | 1 - 2 | 165 | 89 | |
AY118927.1 | - | 2 - 3 | 277 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
