Assay Details
Gene Symbol: Whamy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043234.2 | NP_001036699.1 | 3 - 4 | 1831 | 67 | |
NM_001275506.1 | NP_001262435.1 | 3 - 4 | 1488 | 67 | ||
NM_001275507.1 | NP_001262436.1 | 4 - 5 | 1729 | 67 | ||
NM_001275508.1 | NP_001262437.1 | 4 - 5 | 1738 | 67 | ||
NM_141687.1 | NP_649944.1 | 3 - 4 | 1497 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
