Assay Details
| Gene Symbol: Art4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275516.1 | NP_001262445.1 | 3 - 4 | 1890 | 60 | |
| NM_141706.3 | NP_649963.1 | 3 - 4 | 1890 | 60 | ||
| GenBank mRNA | BT126023.1 | - | 3 - 4 | 1885 | 60 | |
More Information
Set Membership: |
|

Art4
Genomic Map