Assay Details
| Gene Symbol: Fancl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300337.1 | NP_001287266.1 | 1 - 2 | 168 | 71 | |
| NM_141717.3 | NP_649974.1 | 1 - 2 | 168 | 71 | ||
| GenBank mRNA | AY075174.1 | - | 1 - 2 | 145 | 71 | |
More Information
Set Membership: |
|

Fancl
Genomic Map