Assay Details
| Gene Symbol: Bruce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275531.2 | NP_001262460.1 | 29 - 30 | 12863 | 64 | |
| NM_001275532.2 | NP_001262461.1 | 27 - 28 | 12491 | 64 | ||
| NM_001275533.2 | NP_001262462.1 | 28 - 29 | 12560 | 64 | ||
| NM_001275534.2 | NP_001262463.1 | 28 - 29 | 12530 | 64 | ||
| NM_141738.5 | NP_649995.2 | 28 - 29 | 12563 | 64 | ||
| GenBank mRNA | AF517634.1 | - | 28 - 29 | 12560 | 64 | |
More Information
Set Membership: |
|

Bruce
Genomic Map