Assay Details
Gene Symbol: Bruce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275531.2 | NP_001262460.1 | 3 - 4 | 1759 | 69 | |
NM_001275532.2 | NP_001262461.1 | 3 - 4 | 1759 | 69 | ||
NM_001275533.2 | NP_001262462.1 | 3 - 4 | 1759 | 69 | ||
NM_001275534.2 | NP_001262463.1 | 3 - 4 | 1759 | 69 | ||
NM_141738.5 | NP_649995.2 | 3 - 4 | 1759 | 69 | ||
GenBank mRNA | AF517634.1 | - | 3 - 4 | 1756 | 69 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
