Assay Details
Gene Symbol: Mrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141829.3 | NP_650086.2 | 3 - 4 | 822 | 89 | |
GenBank mRNA | BT031028.1 | - | 3 - 4 | 822 | 89 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
