Assay Details
Gene Symbol: Mrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141829.3 | NP_650086.2 | 4 - 5 | 997 | 79 | |
GenBank mRNA | BT031028.1 | - | 4 - 5 | 997 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
