Assay Details
| Gene Symbol: Csk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275547.1 | NP_001262476.1 | 8 - 9 | 3114 | 58 | |
| NM_141840.1 | NP_650097.2 | 8 - 9 | 2370 | 58 | ||
| NM_169414.2 | NP_731607.1 | 8 - 9 | 2806 | 58 | ||
| NM_169415.2 | NP_731608.1 | 8 - 9 | 2234 | 58 | ||
| NM_169416.2 | NP_731609.1 | 7 - 8 | 2454 | 58 | ||
| NM_169418.3 | NP_731611.2 | 9 - 10 | 3375 | 58 | ||
| NM_176461.1 | NP_788638.1 | 8 - 9 | 2228 | 58 | ||
More Information
Set Membership: |
|

Csk
Genomic Map