Assay Details
| Gene Symbol: HisCl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170120.2 | NP_001163591.1 | 2 - 3 | 428 | 84 | |
| NM_141859.4 | NP_650116.2 | 2 - 3 | 428 | 84 | ||
| NM_169429.4 | NP_731632.1 | 2 - 3 | 428 | 84 | ||
| NM_206474.3 | NP_996196.1 | 1 - 2 | 310 | 84 | ||
| GenBank mRNA | AF382401.1 | - | 1 - 2 | 310 | 84 | |
| AF382402.1 | - | 1 - 2 | 310 | 84 | ||
| AF435470.1 | - | 1 - 2 | 112 | 84 | ||
| AF435471.1 | - | 1 - 2 | 112 | 84 | ||
| AY049775.1 | - | 1 - 2 | 264 | 84 | ||
| AY060303.1 | - | 1 - 2 | 264 | 84 | ||
More Information
Set Membership: |
|

HisCl1
Genomic Map