Assay Details
| Gene Symbol: ninaB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142050.3 | NP_650307.2 | 2 - 3 | 691 | 61 | |
| GenBank mRNA | AJ276682.1 | - | 2 - 3 | 555 | 61 | |
| AY121617.1 | - | 2 - 3 | 666 | 61 | ||
More Information
Set Membership: |
|

ninaB
Genomic Map