Assay Details
Gene Symbol: red | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260168.1 | NP_001247097.1 | 2 - 3 | 265 | 86 | |
NM_001300396.1 | NP_001287325.1 | 3 - 4 | 687 | 86 | ||
NM_142095.4 | NP_650352.1 | 2 - 3 | 468 | 86 | ||
NM_169572.3 | NP_731894.1 | 2 - 3 | 149 | 86 | ||
GenBank mRNA | AY051674.1 | - | 2 - 3 | 250 | 86 |

More Information
Set Membership: |
![]() ![]() |
