Assay Details
| Gene Symbol: Gyc88E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043246.4 | NP_001036711.1 | 12 - 13 | 1997 | 108 | |
| NM_001260186.2 | NP_001247115.1 | 11 - 12 | 1903 | 108 | ||
| NM_169616.4 | NP_731974.4 | 11 - 12 | 1903 | 108 | ||
More Information
Set Membership: |
|

Gyc88E
Genomic Map