Assay Details
Gene Symbol: tefu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043247.2 | NP_001036712.1 | 14 - 15 | 3441 | 86 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
